Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343296.1 | NP_001330208.1 | 5 - 6 | 1250 | 68 | |
NM_121385.5 | NP_196886.1 | 6 - 7 | 1478 | 68 | ||
GenBank mRNA | AY029195.1 | - | 6 - 7 | 1403 | 68 | |
BT046176.1 | - | 5 - 6 | 1110 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |