Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121396.4 | NP_196897.1 | 1 - 2 | 480 | 63 | |
GenBank mRNA | AY044331.1 | - | 1 - 2 | 194 | 63 | |
AY058155.1 | - | 1 - 2 | 262 | 63 | ||
AY090376.1 | - | 1 - 2 | 266 | 63 | ||
BT000596.1 | - | 1 - 2 | 194 | 63 | ||
DQ062385.1 | - | 1 - 2 | 194 | 63 | ||
DQ062387.1 | - | 1 - 2 | 194 | 63 | ||
DQ062388.1 | - | 1 - 2 | 194 | 63 | ||
DQ062389.1 | - | 1 - 2 | 194 | 63 | ||
DQ062391.1 | - | 1 - 2 | 194 | 63 | ||
DQ062392.1 | - | 1 - 2 | 194 | 63 | ||
DQ062393.1 | - | 1 - 2 | 194 | 63 | ||
DQ062394.1 | - | 1 - 2 | 194 | 63 | ||
DQ062395.1 | - | 1 - 2 | 194 | 63 | ||
DQ062397.1 | - | 1 - 2 | 194 | 63 | ||
DQ062398.1 | - | 1 - 2 | 194 | 63 | ||
DQ062399.1 | - | 1 - 2 | 194 | 63 | ||
DQ062401.1 | - | 1 - 2 | 194 | 63 | ||
DQ062402.1 | - | 1 - 2 | 194 | 63 | ||
DQ062403.1 | - | 1 - 2 | 194 | 63 | ||
DQ062404.1 | - | 1 - 2 | 194 | 63 | ||
DQ062407.1 | - | 1 - 2 | 194 | 63 | ||
DQ062408.1 | - | 1 - 2 | 194 | 63 | ||
DQ062409.1 | - | 1 - 2 | 194 | 63 | ||
DQ062410.1 | - | 1 - 2 | 194 | 63 | ||
DQ062411.1 | - | 1 - 2 | 194 | 63 | ||
DQ062412.1 | - | 1 - 2 | 194 | 63 | ||
DQ062413.1 | - | 1 - 2 | 194 | 63 | ||
DQ062416.1 | - | 1 - 2 | 194 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |