Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROXY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121410.3 | NP_196911.1 | 1 | 633 | 131 | |
GenBank mRNA | AF361802.1 | - | 1 | 577 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |