Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121413.2 | NP_196914.1 | 3 - 4 | 449 | 94 | |
GenBank mRNA | AK118738.1 | - | 3 - 4 | 338 | 94 | |
AY085446.1 | - | 3 - 4 | 449 | 94 | ||
BT006054.1 | - | 3 - 4 | 238 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |