Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203379.1 | NP_001190308.1 | 4 - 5 | 1891 | 80 | |
NM_001343329.1 | NP_001332743.1 | 5 - 6 | 1672 | 80 | ||
NM_121431.3 | NP_568297.1 | 4 - 5 | 1896 | 80 | ||
GenBank mRNA | AY059785.1 | - | 4 - 5 | 1740 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |