Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343350.1 | NP_001329975.1 | 7 - 8 | 775 | 88 | |
NM_121468.4 | NP_196968.2 | 7 - 8 | 979 | 88 | ||
GenBank mRNA | AY065043.1 | - | 7 - 8 | 887 | 88 | |
BT010466.1 | - | 6 - 7 | 674 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |