Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121499.3 | NP_196999.1 | 2 - 3 | 1000 | 84 | |
GenBank mRNA | DQ029214.1 | - | 2 - 3 | 876 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |