Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GASA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121527.4 | NP_197027.1 | 1 - 2 | 258 | 75 | |
GenBank mRNA | AF360199.1 | - | 1 - 2 | 171 | 75 | |
AY040048.1 | - | 1 - 2 | 57 | 75 | ||
AY057601.1 | - | 1 - 2 | 171 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |