Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343439.1 | NP_001332392.1 | 2 - 3 | 1142 | 93 | |
NM_121623.3 | NP_197122.2 | 2 - 3 | 1142 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |