Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343439.1 | NP_001332392.1 | 5 - 6 | 2170 | 83 | |
NM_121623.3 | NP_197122.2 | 6 - 7 | 2079 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |