Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343440.1 | NP_001330943.1 | 1 - 2 | 622 | 67 | |
NM_001343441.1 | NP_001330942.1 | 1 - 2 | 622 | 67 | ||
NM_001343442.1 | NP_001330941.1 | 1 - 2 | 560 | 67 | ||
NM_121624.4 | NP_197123.3 | 1 - 2 | 73 | 67 | ||
GenBank mRNA | KJ138686.1 | - | 1 - 2 | 73 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |