Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343440.1 | NP_001330943.1 | 2 - 3 | 721 | 91 | |
NM_001343441.1 | NP_001330942.1 | 2 - 3 | 721 | 91 | ||
NM_001343442.1 | NP_001330941.1 | 2 - 3 | 659 | 91 | ||
NM_121624.4 | NP_197123.3 | 2 - 3 | 172 | 91 | ||
GenBank mRNA | KJ138686.1 | - | 2 - 3 | 172 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |