Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLN1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121663.3 | NP_568335.1 | 10 - 11 | 980 | 87 | |
GenBank mRNA | AY059932.1 | - | 10 - 11 | 797 | 87 | |
AY128749.1 | - | 10 - 11 | 754 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |