Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLN1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343456.1 | NP_001331815.1 | 2 - 3 | 213 | 66 | |
NM_121663.3 | NP_568335.1 | 2 - 3 | 347 | 66 | ||
GenBank mRNA | AY059932.1 | - | 2 - 3 | 164 | 66 | |
AY128749.1 | - | 2 - 3 | 121 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |