Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121668.4 | NP_197165.1 | 6 - 7 | 996 | 71 | |
GenBank mRNA | AF428299.1 | - | 6 - 7 | 918 | 71 | |
AY093010.1 | - | 6 - 7 | 879 | 71 | ||
BT006595.1 | - | 6 - 7 | 808 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |