Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121676.4 | NP_568336.1 | 2 - 3 | 343 | 116 | |
GenBank mRNA | AK118603.1 | - | 2 - 3 | 314 | 116 | |
AK175537.1 | - | 2 - 3 | 315 | 116 | ||
AK175798.1 | - | 2 - 3 | 314 | 116 | ||
AK176039.1 | - | 2 - 3 | 315 | 116 | ||
AK176071.1 | - | 2 - 3 | 304 | 116 | ||
AK176708.1 | - | 2 - 3 | 315 | 116 | ||
AK176870.1 | - | 2 - 3 | 314 | 116 | ||
AY085616.1 | - | 2 - 3 | 315 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |