Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343464.1 | NP_001318573.1 | 11 - 12 | 2603 | 72 | |
NM_001343465.1 | NP_001330040.1 | 11 - 12 | 2600 | 72 | ||
NM_121684.4 | NP_197180.1 | 11 - 12 | 2600 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |