Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121688.4 | NP_197184.2 | 2 - 3 | 1698 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |