Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343468.1 | NP_001330126.1 | 4 - 5 | 768 | 83 | |
NM_121691.4 | NP_197187.1 | 4 - 5 | 670 | 83 | ||
GenBank mRNA | AF135454.1 | - | 4 - 5 | 654 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |