Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343468.1 | NP_001330126.1 | 7 - 8 | 1319 | 129 | |
NM_121691.4 | NP_197187.1 | 7 - 8 | 1221 | 129 | ||
GenBank mRNA | AF135454.1 | - | 7 - 8 | 1205 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |