Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121730.4 | NP_197226.2 | 1 - 2 | 170 | 62 | |
GenBank mRNA | AK175921.1 | - | 1 - 2 | 141 | 62 | |
BT010455.1 | - | 1 - 2 | 139 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |