Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121738.2 | NP_197234.1 | 9 - 10 | 1799 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |