Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203398.1 | NP_001190327.1 | 3 - 4 | 1025 | 97 | |
NM_001343523.1 | NP_001330983.1 | 3 - 4 | 1022 | 97 | ||
NM_121787.3 | NP_197283.2 | 2 - 3 | 940 | 97 | ||
GenBank mRNA | AK117910.1 | - | 2 - 3 | 862 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |