Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121817.4 | NP_568360.1 | 1 - 2 | 640 | 69 | |
GenBank mRNA | BT002852.1 | - | 1 - 2 | 451 | 69 | |
BT004434.1 | - | 1 - 2 | 164 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons |