Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121833.7 | NP_197329.5 | 1 - 2 | 684 | 112 | |
GenBank mRNA | AF156783.1 | - | 1 - 2 | 639 | 112 | |
AY099866.1 | - | 1 - 2 | 630 | 112 | ||
BT000329.1 | - | 1 - 2 | 321 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |