Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP1;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203400.1 | NP_001190329.1 | 3 - 4 | 701 | 100 | |
NM_121834.2 | NP_197330.1 | 2 - 3 | 868 | 100 | ||
GenBank mRNA | AK117849.1 | - | 2 - 3 | 626 | 100 | |
BT005263.1 | - | 2 - 3 | 575 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |