Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGSIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121853.4 | NP_197349.2 | 5 - 6 | 1151 | 100 | |
GenBank mRNA | AK118951.1 | - | 5 - 6 | 992 | 100 | |
KJ138680.1 | - | 5 - 6 | 956 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |