Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121863.4 | NP_568364.1 | 8 - 9 | 1001 | 73 | |
GenBank mRNA | AF280057.1 | - | 8 - 9 | 977 | 73 | |
AF360316.1 | - | 8 - 9 | 964 | 73 | ||
AY113888.1 | - | 8 - 9 | 819 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |