Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121873.4 | NP_197369.2 | 1 - 2 | 515 | 138 | |
GenBank mRNA | AB493753.1 | - | 1 - 2 | 311 | 138 | |
BT010768.1 | - | 1 - 2 | 342 | 138 | ||
BT020601.1 | - | 1 - 2 | 311 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |