Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343562.1 | NP_001330131.1 | 3 - 4 | 927 | 89 | |
NM_121873.4 | NP_197369.2 | 3 - 4 | 736 | 89 | ||
GenBank mRNA | AB493753.1 | - | 3 - 4 | 532 | 89 | |
BT010768.1 | - | 3 - 4 | 563 | 89 | ||
BT020601.1 | - | 3 - 4 | 532 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |