Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343577.1 | NP_001330829.1 | 2 | 2186 | 94 | |
NM_121901.2 | NP_197397.1 | 2 | 2104 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |