Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343592.1 | NP_001332355.1 | 6 - 7 | 965 | 65 | |
NM_121923.4 | NP_568370.1 | 6 - 7 | 965 | 65 | ||
GenBank mRNA | AK226388.1 | - | 6 - 7 | 939 | 65 | |
AY050426.1 | - | 6 - 7 | 902 | 65 | ||
AY120692.1 | - | 6 - 7 | 838 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |