Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121927.3 | NP_197423.1 | 12 - 13 | 1362 | 113 | |
GenBank mRNA | AF370503.1 | - | 12 - 13 | 1361 | 113 | |
BT008884.1 | - | 12 - 13 | 1306 | 113 | ||
U72290.1 | - | 12 - 13 | 1319 | 113 | ||
X73367.2 | - | 1319 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |