Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161248.1 | NP_001154720.1 | 12 - 13 | 1721 | 91 | |
NM_121933.4 | NP_197429.1 | 11 - 12 | 1701 | 91 | ||
GenBank mRNA | AY045853.1 | - | 11 - 12 | 1647 | 91 | |
AY117152.1 | - | 11 - 12 | 1560 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |