Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343605.1 | NP_001318599.1 | 1 - 2 | 325 | 155 | |
NM_121952.3 | NP_197448.2 | 1 - 2 | 357 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |