Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128609.4 | NP_180615.1 | 1 - 2 | 352 | 63 | |
GenBank mRNA | AF325042.2 | - | 1 - 2 | 153 | 63 | |
AF375448.1 | - | 1 - 2 | 193 | 63 | ||
AY087402.1 | - | 1 - 2 | 235 | 63 | ||
AY143958.1 | - | 1 - 2 | 94 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |