Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBP-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128632.4 | NP_180637.3 | 3 - 4 | 641 | 99 | |
GenBank mRNA | AK175738.1 | - | 3 - 4 | 562 | 99 | |
AK175821.1 | - | 3 - 4 | 562 | 99 | ||
AK175934.1 | - | 3 - 4 | 562 | 99 | ||
BT022094.1 | - | 3 - 4 | 534 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |