Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCO ARATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084517.1 | NP_001077986.1 | 7 - 8 | 1400 | 157 | |
NM_001336323.1 | NP_001324768.1 | 5 - 6 | 1321 | 157 | ||
NM_128673.5 | NP_565717.2 | 7 - 8 | 1408 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |