Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128707.3 | NP_180708.1 | 2 - 3 | 813 | 115 | |
GenBank mRNA | DQ446585.1 | - | 2 - 3 | 535 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |