Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128723.3 | NP_180724.2 | 3 - 4 | 549 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |