Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CB5-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128831.4 | NP_180831.1 | 2 - 3 | 418 | 110 | |
GenBank mRNA | AK227415.1 | - | 2 - 3 | 184 | 110 | |
AY128405.1 | - | 2 - 3 | 189 | 110 | ||
BT000085.1 | - | 2 - 3 | 155 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |