Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202731.1 | NP_001189660.1 | 14 - 15 | 1767 | 105 | |
NM_128841.5 | NP_565755.1 | 14 - 15 | 1837 | 105 | ||
GenBank mRNA | AJ270305.1 | - | 14 - 15 | 1741 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |