Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATZW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128850.3 | NP_565757.2 | 7 - 8 | 1616 | 107 | |
GenBank mRNA | AY069883.1 | - | 7 - 8 | 1550 | 107 | |
U80984.1 | - | 7 - 8 | 1538 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |