Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336416.1 | NP_001318339.1 | 6 - 7 | 1479 | 75 | |
NM_001336417.1 | NP_001325104.1 | 5 - 6 | 974 | 75 | ||
NM_128853.2 | NP_565758.1 | 6 - 7 | 1467 | 75 | ||
GenBank mRNA | AF138744.1 | - | 6 - 7 | 929 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |