Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336418.1 | NP_001325309.1 | 3 - 4 | 666 | 149 | |
NM_128855.4 | NP_180854.1 | 3 - 4 | 669 | 149 | ||
GenBank mRNA | BT025337.1 | - | 3 - 4 | 550 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |