Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336435.1 | NP_001324208.1 | 1 - 2 | 55 | 73 | |
NM_001336436.1 | NP_001324211.1 | 1 - 2 | 55 | 73 | ||
NM_001336438.1 | NP_001324210.1 | 1 - 2 | 316 | 73 | ||
NM_001336439.1 | NP_001324209.1 | 1 - 2 | 55 | 73 | ||
NM_001336440.1 | NP_001324214.1 | 1 - 2 | 55 | 73 | ||
NM_128883.3 | NP_180882.2 | 1 - 2 | 55 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |