Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336434.1 | NP_001324212.1 | 25 - 26 | 3580 | 110 | |
NM_001336435.1 | NP_001324208.1 | 25 - 26 | 3147 | 110 | ||
NM_001336436.1 | NP_001324211.1 | 25 - 26 | 3147 | 110 | ||
NM_001336437.1 | NP_001324213.1 | 11 - 12 | 1619 | 110 | ||
NM_001336438.1 | NP_001324210.1 | 25 - 26 | 3408 | 110 | ||
NM_001336439.1 | NP_001324209.1 | 25 - 26 | 3147 | 110 | ||
NM_001336440.1 | NP_001324214.1 | 25 - 26 | 3147 | 110 | ||
NM_128883.3 | NP_180882.2 | 25 - 26 | 3147 | 110 | ||
GenBank mRNA | LC177513.1 | - | 24 - 25 | 3093 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |