Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWI3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128921.6 | NP_180919.2 | 3 - 4 | 1040 | 77 | |
GenBank mRNA | BT004085.1 | - | 3 - 4 | 1022 | 77 | |
BT005094.1 | - | 3 - 4 | 989 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |