Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128925.2 | NP_180923.1 | 11 - 12 | 1285 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |