Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336475.1 | NP_001324363.1 | 6 - 7 | 1142 | 73 | |
NM_128946.4 | NP_180942.1 | 7 - 8 | 1386 | 73 | ||
GenBank mRNA | AF336917.1 | - | 7 - 8 | 1021 | 73 | |
AF360313.1 | - | 7 - 8 | 1119 | 73 | ||
AY056365.1 | - | 7 - 8 | 1021 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |